siRNA Details
Gene Symbol: Olr450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000288.1 | NP_001000288.1 | 1 | 856 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000288.1 | NP_001000288.1 | 1 | 856 |