siRNA Details
| Gene Symbol: Olr602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000333.1 | NP_001000333.1 | 1 | 613 | ||
| Gene Symbol: LOC102555599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234492.1 | XP_006234554.1 | 1 | 613 | ||

Olr602
Genomic Map