siRNA Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017253.2 | NP_058949.1 | 3 | 366 | ||
| XM_008763370.1 | XP_008761592.1 | 3 | 379 | |||
| XM_008763371.1 | XP_008761593.1 | 3 | 629 | |||
| XM_008763372.1 | XP_008761594.1 | 4 | 455 | |||
| XM_008763373.1 | XP_008761595.1 | 3 | 558 | |||
| GenBank mRNA | AF165887.1 | - | 1 | 123 | ||
| AJ278701.1 | - | 3 | 360 | |||
| BC087710.1 | - | 3 | 366 | 7315507 | ||
| U35774.1 | - | 3 | 332 | |||

Bcat1
Genomic Map