siRNA Details
| Gene Symbol: Gja8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153465.2 | NP_703195.2 | 1 | 1383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gja8| Gene Symbol: Gja8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153465.2 | NP_703195.2 | 1 | 1383 | ||