siRNA Details
| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053312.1 | NP_445764.1 | 5 | 472 | ||
| GenBank mRNA | AB047915.1 | - | 3 | 230 | ||
| BC101904.1 | - | 5 | 472 | |||
| BC166487.1 | - | 5 | 459 | 7103882 | ||
| CK356543.1 | - | 5 | 458 | |||

Dbt
Genomic Map