siRNA Details
| Gene Symbol: RGD1563136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_215875.5 | XP_215875.4 | 2 | 276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563136| Gene Symbol: RGD1563136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_215875.5 | XP_215875.4 | 2 | 276 | ||