siRNA Details
| Gene Symbol: Alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017260.2 | NP_058956.1 | 5 | 399 | ||
| GenBank mRNA | BC086593.1 | - | 5 | 399 | 7310271 | |
| X52196.1 | - | 5 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alox5ap| Gene Symbol: Alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017260.2 | NP_058956.1 | 5 | 399 | ||
| GenBank mRNA | BC086593.1 | - | 5 | 399 | 7310271 | |
| X52196.1 | - | 5 | 381 | |||