siRNA Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019253.3 | NP_062126.3 | 11 | 1775 | ||
| XM_006229252.2 | XP_006229314.1 | 16 | 2680 | |||
| XM_006229253.2 | XP_006229315.1 | 16 | 2621 | |||
| XM_006229254.2 | XP_006229316.1 | 15 | 2520 | |||
| XM_006229255.2 | XP_006229317.1 | 15 | 2596 | |||
| XM_006229257.2 | XP_006229319.1 | 14 | 2427 | |||
| XM_008759346.1 | XP_008757568.1 | 16 | 2155 | |||
| GenBank mRNA | BC092196.1 | - | 9 | 1140 | 7313460 | |
| FQ211673.1 | - | 16 | 2284 | |||
| S49400.1 | - | 11 | 1775 | |||

Ptpn5
Genomic Map