siRNA Details
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011961.1 | NP_001011961.1 | 4 | 909 | ||
GenBank mRNA | BC090006.1 | - | 4 | 909 | 7323399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011961.1 | NP_001011961.1 | 4 | 909 | ||
GenBank mRNA | BC090006.1 | - | 4 | 909 | 7323399 |