siRNA Details
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011961.1 | NP_001011961.1 | 7 | 1332 | ||
GenBank mRNA | BC090006.1 | - | 7 | 1332 | 7323399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011961.1 | NP_001011961.1 | 7 | 1332 | ||
GenBank mRNA | BC090006.1 | - | 7 | 1332 | 7323399 |