siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198744.2 | NP_942039.1 | 6 | 654 | ||
| XM_006235754.2 | XP_006235816.1 | 7 | 680 | |||
| XM_006235757.2 | XP_006235819.1 | 6 | 640 | |||
| XM_006235760.2 | XP_006235822.1 | 5 | 608 | |||
| XM_008762499.1 | XP_008760721.1 | 7 | 614 | |||
| GenBank mRNA | BC059167.1 | - | 6 | 655 | 6890828 | |

Tpd52l2
Genomic Map