siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198744.2 | NP_942039.1 | 2 | 200 | ||
| XM_006235754.2 | XP_006235816.1 | 2 | 199 | |||
| XM_006235757.2 | XP_006235819.1 | 2 | 219 | |||
| XM_006235759.2 | XP_006235821.1 | 2 | 192 | |||
| XM_006235760.2 | XP_006235822.1 | 2 | 214 | |||
| XM_006235761.2 | XP_006235823.1 | 2 | 197 | |||
| GenBank mRNA | BC059167.1 | - | 2 | 201 | 6890828 | |

Tpd52l2
Genomic Map