siRNA Details
| Gene Symbol: Dlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100531.1 | NP_001094001.1 | 1 | 804 | ||
| XM_008761894.1 | XP_008760116.1 | 1 | 715 | |||
| GenBank mRNA | BC167073.1 | - | 1 | 454 | 8364673 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx1| Gene Symbol: Dlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100531.1 | NP_001094001.1 | 1 | 804 | ||
| XM_008761894.1 | XP_008760116.1 | 1 | 715 | |||
| GenBank mRNA | BC167073.1 | - | 1 | 454 | 8364673 | |