siRNA Details
| Gene Symbol: Arntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024362.2 | NP_077338.2 | 20 | 2290 | ||
| XM_006230029.2 | XP_006230091.1 | 18 | 2048 | |||
| XM_006230030.2 | XP_006230092.1 | 18 | 2045 | |||
| XM_008759716.1 | XP_008757938.1 | 18 | 1988 | |||
| GenBank mRNA | AB012600.1 | - | 15 | 1722 | ||
| AF015953.1 | - | 14 | 1619 | |||
| BC099833.1 | - | 20 | 2290 | 7936079 | ||

Arntl
Genomic Map