siRNA Details
| Gene Symbol: Arntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024362.2 | NP_077338.2 | 14 | 1524 | ||
| XM_006230029.2 | XP_006230091.1 | 12 | 1282 | |||
| XM_006230030.2 | XP_006230092.1 | 12 | 1279 | |||
| XM_008759716.1 | XP_008757938.1 | 12 | 1222 | |||
| GenBank mRNA | AB012600.1 | - | 9 | 956 | ||
| AF015953.1 | - | 8 | 853 | |||
| AF317669.2 | - | 9 | 854 | |||
| BC099833.1 | - | 14 | 1524 | 7936079 | ||

Arntl
Genomic Map