siRNA Details
| Gene Symbol: P2rx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031594.1 | NP_113782.1 | 9 | 1063 | ||
| GenBank mRNA | BC078792.1 | - | 9 | 1004 | 7123247 | |
| U32497.1 | - | 9 | 1040 | |||
| U47031.1 | - | 9 | 993 | |||
| X87763.1 | - | 9 | 1063 | |||
| X91200.1 | - | 9 | 978 | |||
| X93565.1 | - | 9 | 1150 | |||

P2rx4
Genomic Map