siRNA Details
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011966.1 | NP_001011966.1 | 5 | 478 | ||
XM_006235883.2 | XP_006235945.1 | 4 | 539 | |||
XM_006235884.2 | XP_006235946.1 | 4 | 539 | |||
XM_006235885.2 | XP_006235947.1 | 4 | 539 | |||
XM_006235886.2 | XP_006235948.1 | 5 | 526 | |||
XM_006235887.2 | XP_006235949.1 | 5 | 526 | |||
XM_006235888.2 | XP_006235950.1 | 4 | 539 | |||
XM_006235889.2 | XP_006235951.1 | 4 | 539 | |||
GenBank mRNA | BC085349.1 | - | 5 | 478 | 7107899 |
