siRNA Details
| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271045.1 | NP_001257974.1 | 4 | 675 | ||
| NM_001271046.2 | NP_001257975.1 | 4 | 542 | |||
| NM_017288.2 | NP_058984.1 | 4 | 756 | |||
| GenBank mRNA | BC094523.1 | - | 4 | 675 | 7316686 | |
| FQ215379.1 | - | 2 | 333 | |||
| FQ216374.1 | - | 4 | 542 | |||
| M91808.1 | - | 4 | 755 | |||

Scn1b
Genomic Map