siRNA Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181371.2 | NP_852036.1 | 8 | 701 | ||
| XM_006236391.2 | XP_006236453.1 | 7 | 643 | |||
| GenBank mRNA | FJ179400.1 | - | 8 | 667 | ||
| FQ211030.1 | - | 8 | 719 | |||
| FQ214179.1 | - | 8 | 784 | |||
| FQ220456.1 | - | 8 | 757 | |||
| S83436.1 | - | 8 | 701 | |||

Gstk1
Genomic Map