siRNA Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181371.2 | NP_852036.1 | 7, 8 | 663 | ||
| XM_006236391.2 | XP_006236453.1 | 6, 7 | 605 | |||
| GenBank mRNA | FJ179400.1 | - | 7, 8 | 629 | ||
| FQ211030.1 | - | 7, 8 | 681 | |||
| FQ214179.1 | - | 7, 8 | 746 | |||
| FQ220456.1 | - | 7, 8 | 719 | |||
| S83436.1 | - | 7, 8 | 663 | |||

Gstk1
Genomic Map