siRNA Details
| Gene Symbol: Ptprn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031600.1 | NP_113788.1 | 11 | 1739 | ||
| GenBank mRNA | U73458.1 | - | 11 | 1739 | ||
| Z50735.1 | - | 11 | 1713 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprn2| Gene Symbol: Ptprn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031600.1 | NP_113788.1 | 11 | 1739 | ||
| GenBank mRNA | U73458.1 | - | 11 | 1739 | ||
| Z50735.1 | - | 11 | 1713 | |||