siRNA Details
| Gene Symbol: Mad2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106594.1 | NP_001100064.1 | 2 | 300 | ||
| XM_006236619.2 | XP_006236681.1 | 2 | 390 | |||
| XM_006236620.2 | XP_006236682.1 | 2 | 390 | |||
| XM_006236621.2 | XP_006236683.1 | 2 | 390 | |||
| GenBank mRNA | FQ222936.1 | - | 2 | 276 | ||

Mad2l1
Genomic Map