siRNA Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080478.1 | NP_536726.1 | 12 | 1719 | ||
| XM_006229935.1 | XP_006229997.1 | 12 | 2082 | |||
| XM_006229936.2 | XP_006229998.1 | 13 | 1924 | |||
| XM_006229938.2 | XP_006230000.1 | 13 | 2263 | |||
| XM_006229940.2 | XP_006230002.1 | 12 | 1104 | |||
| XM_006229941.2 | XP_006230003.1 | 12 | 1138 | |||
| XM_008759717.1 | XP_008757939.1 | 11 | 1097 | |||
| GenBank mRNA | AF333983.1 | - | 12 | 1719 | ||
| X60469.1 | - | 12 | 1414 | |||

Apbb1
Genomic Map