siRNA Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024982.1 | NP_001020153.1 | 17 | 1288 | ||
| XM_006237022.2 | XP_006237084.1 | 17 | 2268 | |||
| XM_008763190.1 | XP_008761412.1 | 16 | 2265 | |||
| GenBank mRNA | BC091347.1 | - | 17 | 1288 | 7316601 | |
| FQ213359.1 | - | 8 | 541 | |||

Cpne9
Genomic Map