siRNA Details
| Gene Symbol: RGD1311164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106623.1 | NP_001100093.1 | 6, 7 | 805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311164| Gene Symbol: RGD1311164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106623.1 | NP_001100093.1 | 6, 7 | 805 | ||