siRNA Details
| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106636.1 | NP_001100106.1 | 6 | 450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bsdc1| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106636.1 | NP_001100106.1 | 6 | 450 | ||