siRNA Details
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106636.1 | NP_001100106.1 | 11 | 2100 | ||
GenBank mRNA | FQ230812.1 | - | 1 | 312 |
Gene Symbol: Tssk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238925.2 | XP_006238987.1 | 2 | 1876 |
