siRNA Details
| Gene Symbol: Tssk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007650.1 | NP_001007651.1 | 2 | 541 | ||
| XM_006238925.2 | XP_006238987.1 | 2 | 316 | |||
| GenBank mRNA | AY346102.1 | - | 2 | 541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tssk3| Gene Symbol: Tssk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007650.1 | NP_001007651.1 | 2 | 541 | ||
| XM_006238925.2 | XP_006238987.1 | 2 | 316 | |||
| GenBank mRNA | AY346102.1 | - | 2 | 541 | ||