siRNA Details
| Gene Symbol: Atp6v0d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011972.1 | NP_001011972.1 | 6 | 893 | ||
| GenBank mRNA | BC089917.1 | - | 6 | 893 | 7306609 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v0d2| Gene Symbol: Atp6v0d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011972.1 | NP_001011972.1 | 6 | 893 | ||
| GenBank mRNA | BC089917.1 | - | 6 | 893 | 7306609 | |