siRNA Details
| Gene Symbol: Olr847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000403.1 | NP_001000403.1 | 1 | 631 | ||
| Gene Symbol: LOC100912327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238173.2 | XP_006238235.1 | 1 | 715 | ||
| Gene Symbol: LOC103690011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763720.1 | XP_008761942.1 | 1 | 715 | ||

Olr847
Genomic Map