siRNA Details
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106655.2 | NP_001100125.2 | 3, 4 | 473 | ||
GenBank mRNA | BC161979.1 | - | 3, 4 | 467 | 9029956 | |
CV116524.1 | - | 2, 3 | 229 |
