siRNA Details
| Gene Symbol: Mrps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007653.1 | NP_001007654.1 | 5 | 408 | ||
| GenBank mRNA | BC083856.1 | - | 5 | 408 | 7125450 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps15| Gene Symbol: Mrps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007653.1 | NP_001007654.1 | 5 | 408 | ||
| GenBank mRNA | BC083856.1 | - | 5 | 408 | 7125450 | |