siRNA Details
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007653.1 | NP_001007654.1 | 6 | 488 | ||
GenBank mRNA | BC083856.1 | - | 6 | 488 | 7125450 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007653.1 | NP_001007654.1 | 6 | 488 | ||
GenBank mRNA | BC083856.1 | - | 6 | 488 | 7125450 |