siRNA Details
| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106686.1 | NP_001100156.1 | 14 | 1919 | ||
| XM_006238862.2 | XP_006238924.1 | 14 | 2191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ago4| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106686.1 | NP_001100156.1 | 14 | 1919 | ||
| XM_006238862.2 | XP_006238924.1 | 14 | 2191 | |||