siRNA Details
| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008515.1 | NP_001008515.1 | 3 | 421 | ||
| GenBank mRNA | BC086605.1 | - | 3 | 421 | 7316796 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qa| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008515.1 | NP_001008515.1 | 3 | 421 | ||
| GenBank mRNA | BC086605.1 | - | 3 | 421 | 7316796 | |