siRNA Details
Gene Symbol: Ubxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013093.1 | NP_001013111.1 | 2 | 180 | ||
GenBank mRNA | BC079223.1 | - | 2 | 180 | 7132226 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013093.1 | NP_001013111.1 | 2 | 180 | ||
GenBank mRNA | BC079223.1 | - | 2 | 180 | 7132226 |