siRNA Details
| Gene Symbol: Pla2g2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013428.1 | NP_001013446.1 | 3, 4 | 306 | ||
| GenBank mRNA | BC079065.1 | - | 3, 4 | 319 | ||
| BC091221.1 | - | 3, 4 | 306 | 7125035 | ||
| FQ227390.1 | - | 3, 4 | 283 | |||
| FQ231588.1 | - | 3, 4 | 283 | |||
| FQ234103.1 | - | 3, 4 | 283 | |||

Pla2g2d
Genomic Map