siRNA Details
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199096.1 | NP_954527.1 | 3 | 291 | ||
XM_006239179.2 | XP_006239241.1 | 3 | 324 | |||
XM_006239180.2 | XP_006239242.1 | 3 | 325 | |||
GenBank mRNA | BC061826.1 | - | 3 | 291 | 5599028 |
