siRNA Details
| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199096.1 | NP_954527.1 | 6 | 664 | ||
| XM_006239179.2 | XP_006239241.1 | 6 | 697 | |||
| GenBank mRNA | BC061826.1 | - | 6 | 664 | 5599028 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Necap2| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199096.1 | NP_954527.1 | 6 | 664 | ||
| XM_006239179.2 | XP_006239241.1 | 6 | 697 | |||
| GenBank mRNA | BC061826.1 | - | 6 | 664 | 5599028 | |