siRNA Details
| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199096.1 | NP_954527.1 | 4 | 384 | ||
| XM_006239179.2 | XP_006239241.1 | 4 | 417 | |||
| XM_006239180.2 | XP_006239242.1 | 4 | 418 | |||
| GenBank mRNA | BC061826.1 | - | 4 | 384 | 5599028 | |

Necap2
Genomic Map