siRNA Details
| Gene Symbol: RGD1562415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221359.6 | XP_221359.1 | 1 | 128 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562415| Gene Symbol: RGD1562415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221359.6 | XP_221359.1 | 1 | 128 | ||