siRNA Details
| Gene Symbol: Ypel5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035221.1 | NP_001030298.1 | 433 | |||
| XM_008764508.1 | XP_008762730.1 | 640 | ||||
| XM_008764509.1 | XP_008762731.1 | 888 | ||||
| XM_008764510.1 | XP_008762732.1 | 839 | ||||
| GenBank mRNA | CB584328.1 | - | 444 | |||
| FQ229713.1 | - | 400 | ||||

Ypel5
Genomic Map