siRNA Details
| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013944.1 | NP_001013966.1 | 10 | 944 | ||
| XM_006240231.1 | XP_006240293.1 | 10 | 1033 | |||
| GenBank mRNA | BC079166.1 | - | 10 | 944 | 7129668 | |
| FQ213708.1 | - | 10 | 957 | |||

Ppp1r36
Genomic Map