siRNA Details
Gene Symbol: RGD1309028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291359.1 | NP_001278288.1 | 2 | 974 | ||
XM_006240509.2 | XP_006240571.1 | 2 | 949 | |||
XM_008764763.1 | XP_008762985.1 | 2 | 1996 |
Gene Symbol: Fam181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240515.2 | XP_006240577.1 | 2 | 937 | ||
XM_006240516.2 | XP_006240578.1 | 2 | 1157 | |||
XM_008764905.1 | XP_008763127.1 | 2 | 1042 |
