siRNA Details
| Gene Symbol: Uxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006982.1 | NP_001006983.1 | 2, 3 | 196 | ||
| XM_006256611.2 | XP_006256673.1 | 3, 4 | 253 | |||
| GenBank mRNA | BC082752.1 | - | 2, 3 | 196 | 7303683 | |
| FQ210799.1 | - | 3, 4 | 241 | |||
| FQ221998.1 | - | 3, 4 | 244 | |||

Uxt
Genomic Map