siRNA Details
| Gene Symbol: Yeats4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127527.2 | NP_001120999.1 | 3, 4 | 259 | ||
| GenBank mRNA | BC166753.1 | - | 3, 4 | 259 | 5623659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yeats4| Gene Symbol: Yeats4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127527.2 | NP_001120999.1 | 3, 4 | 259 | ||
| GenBank mRNA | BC166753.1 | - | 3, 4 | 259 | 5623659 | |