siRNA Details
| Gene Symbol: Eef1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013104.1 | NP_001013122.1 | 1 | 502 | ||
| XM_008765549.1 | XP_008763771.1 | 1 | 2563 | |||
| XM_008765550.1 | XP_008763772.1 | 1 | 627 | |||
| XM_008765551.1 | XP_008763773.1 | 3 | 616 | |||
| XM_008765552.1 | XP_008763774.1 | 3 | 597 | |||
| XM_008765553.1 | XP_008763775.1 | 1 | 516 | |||
| XM_008765554.1 | XP_008763776.1 | 1 | 627 | |||
| XM_008765555.1 | XP_008763777.1 | 1 | 627 | |||
| XM_008765556.1 | XP_008763778.1 | 1 | 627 | |||
| GenBank mRNA | BC079391.1 | - | 1 | 502 | 7111852 | |

Eef1d
Genomic Map