siRNA Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011993.1 | NP_001011993.1 | 3 | 280 | ||
| GenBank mRNA | BC087166.1 | - | 3 | 280 | 7135468 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pycrl| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011993.1 | NP_001011993.1 | 3 | 280 | ||
| GenBank mRNA | BC087166.1 | - | 3 | 280 | 7135468 | |