siRNA Details
| Gene Symbol: Exosc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134860.1 | NP_001128332.1 | 3 | 717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exosc4| Gene Symbol: Exosc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134860.1 | NP_001128332.1 | 3 | 717 | ||