siRNA Details
| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130492.1 | NP_001123964.1 | 7 | 529 | ||
| GenBank mRNA | BC168742.1 | - | 7 | 475 | 6919204 | |
| FQ225519.1 | - | 7 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps28| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130492.1 | NP_001123964.1 | 7 | 529 | ||
| GenBank mRNA | BC168742.1 | - | 7 | 475 | 6919204 | |
| FQ225519.1 | - | 7 | 496 | |||